Product Details

SNP ID
rs169790
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:11714058 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCATTGCCTTGGTTCTGACACTAA[A/G]TAACCCTTTTGCCTTTTGCAGGTAA
Phenotype
MIM: 614348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADTRP PubMed Links

Gene Details

Gene
ADTRP
Gene Name
androgen dependent TFPI regulating protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143948.1 1455 UTR 3 NP_001137420.1
NM_032744.3 1455 UTR 3 NP_116133.1
XM_005249454.3 1455 UTR 3 XP_005249511.1
XM_011514956.1 1455 UTR 3 XP_011513258.1
XM_011514958.2 1455 Intron XP_011513260.1
XM_011514959.1 1455 Intron XP_011513261.1
XM_017011377.1 1455 UTR 3 XP_016866866.1
Gene
LOC340184
Gene Name
uncharacterized LOC340184
There are no transcripts associated with this gene.

View Full Product Details