Product Details

SNP ID
rs723796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:4030914 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTTGTGAGGATGTGTTTATACTAT[C/T]ACCATCATCAGTATCAGCTACCAAT
Phenotype
MIM: 602338
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRPF4B PubMed Links

Gene Details

Gene
PRPF4B
Gene Name
pre-mRNA processing factor 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003913.4 Intron NP_003904.3
XM_011514970.2 Intron XP_011513272.1
XM_017011410.1 Intron XP_016866899.1
XM_017011411.1 Intron XP_016866900.1
XM_017011412.1 Intron XP_016866901.1
XM_017011413.1 Intron XP_016866902.1
XM_017011414.1 Intron XP_016866903.1
XM_017011415.1 Intron XP_016866904.1
XM_017011416.1 Intron XP_016866905.1
XM_017011417.1 Intron XP_016866906.1
XM_017011418.1 Intron XP_016866907.1
XM_017011419.1 Intron XP_016866908.1
XM_017011420.1 Intron XP_016866909.1
XM_017011421.1 Intron XP_016866910.1
XM_017011422.1 Intron XP_016866911.1
XM_017011423.1 Intron XP_016866912.1
XM_017011424.1 Intron XP_016866913.1
XM_017011425.1 Intron XP_016866914.1
XM_017011426.1 Intron XP_016866915.1
XM_017011427.1 Intron XP_016866916.1
XM_017011428.1 Intron XP_016866917.1
XM_017011429.1 Intron XP_016866918.1
XM_017011430.1 Intron XP_016866919.1
XM_017011431.1 Intron XP_016866920.1
XM_017011432.1 Intron XP_016866921.1
XM_017011433.1 Intron XP_016866922.1
XM_017011434.1 Intron XP_016866923.1
XM_017011435.1 Intron XP_016866924.1

View Full Product Details