Product Details

SNP ID
rs442758
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:32587909 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGTTGGGTAATCCTATACTATC[A/T]TCCTAATGAACTTCAAAAACTCAAG
Phenotype
MIM: 605333
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDS5B PubMed Links

Gene Details

Gene
PDS5B
Gene Name
PDS5 cohesin associated factor B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015032.3 Intron NP_055847.1
XM_005266298.4 Intron XP_005266355.1
XM_011534999.2 Intron XP_011533301.1
XM_011535000.2 Intron XP_011533302.1
XM_011535002.2 Intron XP_011533304.1
XM_017020448.1 Intron XP_016875937.1
XM_017020449.1 Intron XP_016875938.1
XM_017020450.1 Intron XP_016875939.1
XM_017020451.1 Intron XP_016875940.1
XM_017020452.1 Intron XP_016875941.1
XM_017020453.1 Intron XP_016875942.1

View Full Product Details