Product Details

SNP ID
rs291101
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:206932463 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATCCCAGGGACTCACCGACGTT[C/T]TTCCTGTGCCGGGCTCTGGCCACCC
Phenotype
MIM: 173880
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PIGR PubMed Links

Gene Details

Gene
PIGR
Gene Name
polymeric immunoglobulin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002644.3 2185 Silent Mutation AAA,AAG K667K NP_002635.2
XM_011509629.1 2185 Silent Mutation AAA,AAG K667K XP_011507931.1

View Full Product Details