Product Details
- SNP ID
-
rs607159
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:88722623 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCTGATCATGTTGCCCATCCTTCA[A/G]AAAGTCGCATGCGCTTGACTGAGGG
- Phenotype
-
MIM: 600662
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MEF2C
PubMed Links
Gene Details
- Gene
- MEF2C
- Gene Name
- myocyte enhancer factor 2C
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001131005.2 |
1518 |
Missense Mutation |
TCT,TTT |
S458F |
NP_001124477.1 |
NM_001193347.1 |
1518 |
Missense Mutation |
TCT,TTT |
S478F |
NP_001180276.1 |
NM_001193348.1 |
1518 |
Missense Mutation |
TCT,TTT |
S412F |
NP_001180277.1 |
NM_001193349.1 |
1518 |
Missense Mutation |
TCT,TTT |
S388F |
NP_001180278.1 |
NM_001193350.1 |
1518 |
Missense Mutation |
TCT,TTT |
S468F |
NP_001180279.1 |
NM_001308002.1 |
1518 |
Missense Mutation |
TCT,TTT |
S460F |
NP_001294931.1 |
NM_002397.4 |
1518 |
Missense Mutation |
TCT,TTT |
S468F |
NP_002388.2 |
XM_005248511.2 |
1518 |
Missense Mutation |
TCT,TTT |
S468F |
XP_005248568.1 |
XM_006714619.2 |
1518 |
Missense Mutation |
TCT,TTT |
S468F |
XP_006714682.1 |
XM_006714625.3 |
1518 |
Missense Mutation |
TCT,TTT |
S478F |
XP_006714688.1 |
XM_011543396.2 |
1518 |
Missense Mutation |
TCT,TTT |
S468F |
XP_011541698.1 |
XM_011543397.2 |
1518 |
Missense Mutation |
TCT,TTT |
S466F |
XP_011541699.1 |
XM_011543400.1 |
1518 |
Missense Mutation |
TCT,TTT |
S436F |
XP_011541702.1 |
XM_011543401.1 |
1518 |
Missense Mutation |
TCT,TTT |
S428F |
XP_011541703.1 |
XM_017009475.1 |
1518 |
Missense Mutation |
TCT,TTT |
S466F |
XP_016864964.1 |
XM_017009476.1 |
1518 |
Missense Mutation |
TCT,TTT |
S460F |
XP_016864965.1 |
XM_017009477.1 |
1518 |
Missense Mutation |
TCT,TTT |
S460F |
XP_016864966.1 |
XM_017009478.1 |
1518 |
Missense Mutation |
TCT,TTT |
S458F |
XP_016864967.1 |
XM_017009479.1 |
1518 |
Missense Mutation |
TCT,TTT |
S436F |
XP_016864968.1 |
XM_017009480.1 |
1518 |
Missense Mutation |
TCT,TTT |
S436F |
XP_016864969.1 |
XM_017009481.1 |
1518 |
Missense Mutation |
TCT,TTT |
S436F |
XP_016864970.1 |
XM_017009482.1 |
1518 |
Missense Mutation |
TCT,TTT |
S428F |
XP_016864971.1 |
XM_017009483.1 |
1518 |
Missense Mutation |
TCT,TTT |
S426F |
XP_016864972.1 |
View Full Product Details