Product Details

SNP ID
rs335437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:177513273 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCGGTTCCCACAAGGTGGACCCC[T/C]GGCTCCAATCAGCTTCAGGGAGACT
Phenotype
MIM: 608170 MIM: 611435 MIM: 615813
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
DDX41 PubMed Links

Gene Details

Gene
DDX41
Gene Name
DEAD-box helicase 41
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321732.1 Intron NP_001308661.1
NM_001321830.1 Intron NP_001308759.1
NM_016222.3 Intron NP_057306.2
Gene
DOK3
Gene Name
docking protein 3
There are no transcripts associated with this gene.

Gene
FAM193B
Gene Name
family with sequence similarity 193 member B
There are no transcripts associated with this gene.

View Full Product Details