Product Details

SNP ID
rs43207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:65629429 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCCTGTTGGTAGGCAGGGTAT[C/T]CCTACCATATCTTATTCTTTGCAAT
Phenotype
MIM: 601747
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC13 PubMed Links

Gene Details

Gene
TRAPPC13
Gene Name
trafficking protein particle complex 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093755.1 Intron NP_001087224.1
NM_001093756.1 Intron NP_001087225.1
NM_001243737.1 Intron NP_001230666.1
NM_024941.3 Intron NP_079217.2
XM_011543652.2 Intron XP_011541954.1
XM_017009890.1 Intron XP_016865379.1
Gene
TRIM23
Gene Name
tripartite motif containing 23
There are no transcripts associated with this gene.

View Full Product Details