Product Details

SNP ID
rs30491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134115522 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAGGCGGCAGAACCCAGGGGTGG[A/G]GAGTGGGGGGCGGGCTTCCCGGGCG
Phenotype
MIM: 189908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCF7 PubMed Links

Gene Details

Gene
TCF7
Gene Name
transcription factor 7 (T-cell specific, HMG-box)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134851.2 Intron NP_001128323.2
NM_003202.3 Intron NP_003193.2
NM_201632.3 Intron NP_963963.1
NM_201634.3 Intron NP_963965.1
NM_213648.3 Intron NP_998813.1
XM_006714678.3 Intron XP_006714741.1
XM_006714679.3 Intron XP_006714742.1
XM_006714680.3 Intron XP_006714743.1
XM_006714682.2 Intron XP_006714745.1
XM_006714684.2 Intron XP_006714747.1
XM_006714685.3 Intron XP_006714748.1
XM_006714686.3 Intron XP_006714749.1
XM_006714687.3 Intron XP_006714750.1
XM_011543604.2 Intron XP_011541906.1
XM_011543606.1 Intron XP_011541908.1
XM_011543607.2 Intron XP_011541909.1
XM_011543608.2 Intron XP_011541910.1
XM_011543609.2 Intron XP_011541911.1
XM_011543610.2 Intron XP_011541912.1
XM_011543611.2 Intron XP_011541913.1
XM_011543612.2 Intron XP_011541914.1
XM_011543613.2 Intron XP_011541915.1
XM_017009789.1 Intron XP_016865278.1
XM_017009790.1 Intron XP_016865279.1

View Full Product Details