Product Details

SNP ID
rs462469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:132300818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCATCAGGCAGCAGTTGAGCA[C/G]GAACGCAAGCAGGGACTGGTGGGAC
Phenotype
MIM: 604190
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC22A4 PubMed Links

Gene Details

Gene
SLC22A4
Gene Name
solute carrier family 22 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003059.2 Intron NP_003050.2
XM_006714675.3 Intron XP_006714738.1
XM_011543589.2 Intron XP_011541891.1
XM_017009776.1 Intron XP_016865265.1

View Full Product Details