Product Details
- SNP ID
-
rs564196
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:48873325 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCCCAGTGATGGGCCTCCTCAGCC[G/A]CGCCTGGAGCCGCCTGAGGGGCCTG
- Phenotype
-
MIM: 607769
MIM: 611048
MIM: 602309
- Polymorphism
- G/A, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PLEKHA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3786734] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PLEKHA4
- Gene Name
- pleckstrin homology domain containing A4
There are no transcripts associated with this gene.
- Gene
- PPP1R15A
- Gene Name
- protein phosphatase 1 regulatory subunit 15A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014330.3 |
361 |
Missense Mutation |
CAC,CGC |
H31R |
NP_055145.3 |
- Gene
- TULP2
- Gene Name
- tubby like protein 2
There are no transcripts associated with this gene.
View Full Product Details