Product Details
- SNP ID
-
rs418560
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:31121650 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAACTTTTTTGTACAATTGCATA[G/T]ACGTGTAAAACCTGCCATTGTTAAC
- Phenotype
-
MIM: 300377
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DMD
PubMed Links
Gene Details
- Gene
- DMD
- Gene Name
- dystrophin
View Full Product Details