Product Details

SNP ID
rs404401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:31121788 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTACTGTCTAATCCTCTTTGTTGT[A/G]TGAATATTATAAAAACCATGCGGGA
Phenotype
MIM: 300377
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DMD PubMed Links

Gene Details

Gene
DMD
Gene Name
dystrophin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000109.3 4488 UTR 3 NP_000100.2
NM_004006.2 4488 UTR 3 NP_003997.1
NM_004009.3 4488 UTR 3 NP_004000.1
NM_004010.3 4488 UTR 3 NP_004001.1
NM_004011.3 4488 UTR 3 NP_004002.2
NM_004012.3 4488 UTR 3 NP_004003.1
NM_004013.2 4488 UTR 3 NP_004004.1
NM_004014.2 4488 UTR 3 NP_004005.1
NM_004015.2 4488 UTR 3 NP_004006.1
NM_004016.2 4488 UTR 3 NP_004007.1
NM_004017.2 4488 UTR 3 NP_004008.1
NM_004018.2 4488 UTR 3 NP_004009.1
NM_004019.2 4488 Intron NP_004010.1
NM_004020.3 4488 UTR 3 NP_004011.2
NM_004021.2 4488 UTR 3 NP_004012.1
NM_004022.2 4488 UTR 3 NP_004013.1
NM_004023.2 4488 UTR 3 NP_004014.1
XM_006724468.2 4488 UTR 3 XP_006724531.1
XM_006724469.3 4488 UTR 3 XP_006724532.1
XM_006724470.3 4488 UTR 3 XP_006724533.1
XM_006724473.2 4488 UTR 3 XP_006724536.1
XM_006724474.3 4488 UTR 3 XP_006724537.1
XM_006724475.2 4488 UTR 3 XP_006724538.1
XM_011545467.1 4488 UTR 3 XP_011543769.1
XM_011545468.2 4488 Intron XP_011543770.1
XM_011545469.1 4488 Intron XP_011543771.1
XM_017029328.1 4488 UTR 3 XP_016884817.1
XM_017029329.1 4488 Intron XP_016884818.1
XM_017029330.1 4488 Intron XP_016884819.1
XM_017029331.1 4488 UTR 3 XP_016884820.1

View Full Product Details