Product Details

SNP ID
rs129922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133665008 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGAGCAGGTTTCCCTATCTTAGG[A/C]AGAAACCTCCGTTACCCCACTTGGG
Phenotype
MIM: 609312 MIM: 604455
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DBH PubMed Links

Gene Details

Gene
DBH
Gene Name
dopamine beta-hydroxylase
There are no transcripts associated with this gene.

Gene
DBH-AS1
Gene Name
DBH antisense RNA 1
There are no transcripts associated with this gene.

Gene
SARDH
Gene Name
sarcosine dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134707.1 Intron NP_001128179.1
NM_007101.3 Intron NP_009032.2
XM_017014367.1 Intron XP_016869856.1
XM_017014368.1 Intron XP_016869857.1

View Full Product Details