Product Details

SNP ID
rs142967670
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12891965 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCTCGCATCCTGTTGGCCAAT[C/T]GCAACGAAGGTGGGCGGGCTGGTGG
Phenotype
MIM: 608801 MIM: 600599 MIM: 611487
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GCDH PubMed Links

Gene Details

Gene
GCDH
Gene Name
glutaryl-CoA dehydrogenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000159.3 339 Missense Mutation CGC,TGC R88C NP_000150.1
NM_013976.3 339 Missense Mutation CGC,TGC R88C NP_039663.1
XM_006722721.2 339 Missense Mutation CGC,TGC R88C XP_006722784.1
XM_011527899.2 339 Missense Mutation CGC,TGC R88C XP_011526201.1
XM_011527900.2 339 Missense Mutation CGC,TGC R88C XP_011526202.1
XM_017026580.1 339 Missense Mutation CGC,TGC R88C XP_016882069.1
Gene
KLF1
Gene Name
Kruppel like factor 1
There are no transcripts associated with this gene.

Gene
SYCE2
Gene Name
synaptonemal complex central element protein 2
There are no transcripts associated with this gene.

View Full Product Details