Product Details

SNP ID
rs2294076
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:156366 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCCGCTCCTCCAGGGACTTGTCTTC[G/A]GTCTTGTCCACCTGTTGGACGGGAC
Phenotype
MIM: 604568
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
DOC2B PubMed Links

Gene Details

Gene
DOC2B
Gene Name
double C2 domain beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003585.4 928 Silent Mutation ACC,ACT T259T NP_003576.2
XM_011535363.2 928 Silent Mutation ACC,ACT T234T XP_011533665.1
XM_017025227.1 928 Intron XP_016880716.1

View Full Product Details