Product Details

SNP ID
rs12459852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21753049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTTTCAGTCACTCTTGAAGT[A/C]ACTCTTGACTGAAGCAAAAAATTAA
Phenotype
MIM: 603982
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC641367 PubMed Links

Gene Details

Gene
LOC641367
Gene Name
cyclin Y like 1 pseudogene
There are no transcripts associated with this gene.

Gene
ZNF100
Gene Name
zinc finger protein 100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173531.3 Intron NP_775802.2

View Full Product Details