Product Details

SNP ID
rs34559546
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:202943592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACTGAAATAGTATGACACTGGG[A/G]GTTGAGAACTGAGCTTTGAAATTGA
Phenotype
MIM: 607945
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADIPOR1 PubMed Links

Gene Details

Gene
ADIPOR1
Gene Name
adiponectin receptor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290553.1 Intron NP_001277482.1
NM_001290557.1 Intron NP_001277486.1
NM_001290629.1 Intron NP_001277558.1
NM_015999.5 Intron NP_057083.2
XM_017001407.1 Intron XP_016856896.1

View Full Product Details