Product Details

SNP ID
rs5934125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:13661837 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAAAGTCATAAATCATCTGGATC[A/G]TTGAGTCTGAACCTTTCCCTTGTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC107985657 PubMed Links

Gene Details

Gene
LOC107985657
Gene Name
uncharacterized LOC107985657
There are no transcripts associated with this gene.

Gene
TCEANC
Gene Name
transcription elongation factor A N-terminal and central domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297563.1 Intron NP_001284492.1
NM_001297564.1 Intron NP_001284493.1
NM_152634.3 Intron NP_689847.2
XM_017029316.1 Intron XP_016884805.1
XM_017029317.1 Intron XP_016884806.1
XM_017029318.1 Intron XP_016884807.1
XM_017029319.1 Intron XP_016884808.1

View Full Product Details