Product Details

SNP ID
rs6948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:184627976 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGAGCCTGCCTCCCGGGCTGAG[G/T]GCTCAGCTCTGGCCTCCGGCTGGAT
Phenotype
MIM: 600636
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CASP3 PubMed Links

Gene Details

Gene
CASP3
Gene Name
caspase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004346.3 2393 UTR 3 NP_004337.2
NM_032991.2 2393 UTR 3 NP_116786.1
XM_011532301.1 2393 UTR 3 XP_011530603.1
Gene
LVCAT8
Gene Name
liver cancer-associated transcript 8
There are no transcripts associated with this gene.

View Full Product Details