Product Details

SNP ID
rs1949755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:104504493 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGACATCTGGTACTCCTCTTCTGC[C/G]CTCTCTCCAATGCTGGCAAACTTTG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
OR13F1 PubMed Links

Gene Details

Gene
OR13F1
Gene Name
olfactory receptor family 13 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004485.1 231 Silent Mutation GCC,GCG A77A NP_001004485.1

View Full Product Details