Product Details

SNP ID
rs621878
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12962871 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCTTTTTTTTTTGTGATCAGTT[G/T]GCTTAAGAGAGATTACTTATATAAC
Phenotype
MIM: 609263
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEH1L PubMed Links

Gene Details

Gene
SEH1L
Gene Name
SEH1 like nucleoporin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013437.1 Intron NP_001013455.1
NM_031216.3 Intron NP_112493.2
XM_005258152.4 Intron XP_005258209.1
XM_011525742.2 Intron XP_011524044.1
XM_011525743.2 Intron XP_011524045.1
XM_011525744.1 Intron XP_011524046.1
XM_011525745.2 Intron XP_011524047.1
XM_011525746.2 Intron XP_011524048.1
XM_017026025.1 Intron XP_016881514.1
XM_017026026.1 Intron XP_016881515.1
XM_017026027.1 Intron XP_016881516.1

View Full Product Details