Product Details

SNP ID
rs692270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:49852448 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTTGCACTTGCTAGAAATCTGAT[C/T]TGTGCATGGCCTAGCCCTTTAGGGA
Phenotype
MIM: 611108 MIM: 608498
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VWC2 PubMed Links

Gene Details

Gene
VWC2
Gene Name
von Willebrand factor C domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198570.4 Intron NP_940972.2
Gene
ZPBP
Gene Name
zona pellucida binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159878.1 Intron NP_001153350.1
NM_007009.2 Intron NP_008940.2
XM_011515095.2 Intron XP_011513397.1
XM_011515096.2 Intron XP_011513398.1
XM_011515097.1 Intron XP_011513399.1
XM_011515100.2 Intron XP_011513402.1
XM_011515101.2 Intron XP_011513403.1
XM_011515102.2 Intron XP_011513404.1
XM_011515103.1 Intron XP_011513405.1
XM_017011707.1 Intron XP_016867196.1

View Full Product Details