Product Details
- SNP ID
-
rs729832
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:75135547 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTGGTATCTTGCCTGGTGCTTCCA[A/G]AGGGAGGGAGAAGGGGAGGCGAACA
- Phenotype
-
MIM: 604425
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LHX8
PubMed Links
Gene Details
- Gene
- LHX8
- Gene Name
- LIM homeobox 8
View Full Product Details