Assay Design
Gene Transcripts
- Gene Symbol
- spe-8
- Entrez Gene ID
-
171606
- Gene Name
- Spermatocyte protein spe-8
- Gene Aliases
-
CELE_F53G12.6
- Chromosome Location
-
Chr.I: 118093 - 120901 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_058279.2 |
NP_490680.1 |
4 - 5 |
1089 |
75 |
View Full Product Details