Assay Design
Gene Transcripts
- Gene Symbol
- atf-4
- Entrez Gene ID
-
181576
- Gene Name
- BZIP domain-containing protein
- Gene Aliases
-
CELE_T04C10.4
- Chromosome Location
-
Chr.X: 14809156 - 14810849 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078055.6 |
NP_510456.1 |
3 - 4 |
802 |
89 |
View Full Product Details