Assay Design
Gene Transcripts
- Gene Symbol
- daf-6
- Entrez Gene ID
-
181584
- Gene Name
- SSD domain-containing protein
- Gene Aliases
-
CELE_F31F6.5
- Chromosome Location
-
Chr.X: 14888488 - 14894333 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078071.7 |
NP_510472.2 |
2 - 3 |
423 |
145 |
View Full Product Details