Assay Design
Gene Transcripts
- Gene Symbol
- mrp-5
- Entrez Gene ID
-
181587
- Gene Name
- Multidrug resistance-associated protein 5
- Gene Aliases
-
CELE_F14F4.3
- Chromosome Location
-
Chr.X: 14938502 - 14946319 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078078.3 |
NP_510479.1 |
3 - 4 |
349 |
119 |
View Full Product Details