Assay Design
Gene Transcripts
- Gene Symbol
- flr-4
- Entrez Gene ID
-
181604
- Gene Name
- Serine/threonine-protein kinase flr-4
- Gene Aliases
-
CELE_F09B12.6
- Chromosome Location
-
Chr.X: 15089397 - 15093585 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078107.6 |
NP_510508.1 |
7 - 8 |
1086 |
70 |
GenBank mRNA |
AB012700.1 |
- |
7 - 8 |
1108 |
70 |
View Full Product Details