Assay Design
Gene Transcripts
- Gene Symbol
- gpn-1
- Entrez Gene ID
-
181658
- Gene Name
- GlyPicaN
- Gene Aliases
-
CELE_F59D12.4
- Chromosome Location
-
Chr.X: 15681693 - 15687477 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078181.4 |
NP_510582.1 |
8 - 9 |
1541 |
109 |
View Full Product Details