Assay Design
Gene Transcripts
- Gene Symbol
- lin-15B
- Entrez Gene ID
-
181662
- Gene Name
- Protein lin-15B
- Gene Aliases
-
CELE_ZK662.4
- Chromosome Location
-
Chr.X: 15726129 - 15731861 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078186.7 |
NP_510587.1 |
6 - 7 |
3194 |
123 |
GenBank mRNA |
U10412.1 |
- |
6 - 7 |
3216 |
123 |
View Full Product Details