Assay Design
Gene Transcripts
- Gene Symbol
- csb-1
- Entrez Gene ID
-
181674
- Gene Name
- human CSB (Cockayne Syndrome B) homolog
- Gene Aliases
-
CELE_F53H4.1
- Chromosome Location
-
Chr.X: 15861294 - 15865918 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078206.5 |
NP_510607.2 |
8 - 9 |
2379 |
85 |
View Full Product Details