Assay Design
Gene Transcripts
- Gene Symbol
- mua-6
- Entrez Gene ID
-
181698
- Gene Name
- Intermediate filament protein ifa-2
- Gene Aliases
-
CELE_W10G6.3
- Chromosome Location
-
Chr.X: 16262289 - 16265686 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078247.9 |
NP_510648.1 |
4 - 5 |
1581 |
73 |
View Full Product Details