Assay Design



Gene Transcripts

Gene Symbol
nhr-28
Entrez Gene ID
181705
Gene Name
protein-coding
Gene Aliases
CELE_C11G6.4
Chromosome Location
Chr.X: 16352477 - 16359654 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029217.6 NP_001024388.1 8 - 9 783 115
NM_001029218.7 NP_001024389.1 5 - 6 837 115
GenBank mRNA AY305831.1 - 6 - 7 589 115
AY305832.1 - 6 - 7 579 115

View Full Product Details