Assay Design
Gene Transcripts
- Gene Symbol
- cas-1
- Entrez Gene ID
-
181729
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F41G4.2
- Chromosome Location
-
Chr.X: 16807455 - 16815478 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078312.6 |
NP_510713.1 |
2 - 3 |
209 |
65 |
View Full Product Details