Assay Design



Gene Transcripts

Gene Symbol
cas-1
Entrez Gene ID
181729
Gene Name
protein-coding
Gene Aliases
CELE_F41G4.2
Chromosome Location
Chr.X: 16807455 - 16815478 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373613.3 NP_001360035.1 3 - 4 3371 76
NM_078312.6 NP_510713.1 4 - 5 565 76

View Full Product Details