Assay Design



Gene Transcripts

Gene Symbol
sto-5
Entrez Gene ID
181730
Gene Name
protein-coding
Gene Aliases
CELE_F41G4.3
Chromosome Location
Chr.X: 16816260 - 16839975 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001368561.3 NP_001355390.1 4 - 5 976 85
NM_001368562.3 NP_001355391.1 4 - 5 592 85
NM_001368563.4 NP_001355544.1 3 - 4 226 85
NM_001383696.1 5 - 6 912 85
NM_001383697.1 4 - 5 345 85

View Full Product Details