Assay Design



Gene Transcripts

Gene Symbol
gcy-11
Entrez Gene ID
181745
Gene Name
Receptor-type guanylate cyclase gcy-11
Gene Aliases
CELE_C30G4.3
Chromosome Location
Chr.X: 17046241 - 17055581 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001373625.3 NP_001359843.1 14 - 15 1781 65

View Full Product Details