Assay Design
Gene Transcripts
- Gene Symbol
- crb-1
- Entrez Gene ID
-
181771
- Gene Name
- Drosophila CRumBs homolog
- Gene Aliases
-
CELE_F11C7.4
- Chromosome Location
-
Chr.X: 17402532 - 17414014 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078421.6 |
NP_510822.1 |
26 - 27 |
4920 |
80 |
View Full Product Details