Assay Design
Gene Transcripts
- Gene Symbol
- cgt-2
- Entrez Gene ID
-
181790
- Gene Name
- Ceramide glucosyltransferase 2
- Gene Aliases
-
CELE_F20B4.6
- Chromosome Location
-
Chr.X: 17695232 - 17701744 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078456.5 |
NP_510857.2 |
5 - 6 |
1120 |
140 |
GenBank mRNA |
AF364401.1 |
- |
5 - 6 |
1124 |
140 |
View Full Product Details