Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306342.3 | NP_001293271.1 | 5 - 6 | 1234 | 80 |
NM_001306343.3 | NP_001293272.1 | 5 - 6 | 1228 | 80 | |
NM_170794.7 | NP_740775.1 | 9 - 10 | 2111 | 80 | |
NM_171831.4 | NP_740776.1 | 9 - 10 | 2105 | 80 |