Assay Design



Gene Transcripts

Gene Symbol
pqn-44
Entrez Gene ID
172187
Gene Name
protein-coding
Gene Aliases
CELE_F55A12.9
Chromosome Location
Chr.I: 5372851 - 5394460 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059179.3 NP_491580.1 4 - 5 874 126
NM_059180.2 NP_491581.1 4 - 5 753 126
NM_170865.5 NP_740852.1 5 - 6 772 126
NM_182025.7 NP_871825.1 4 - 5 659 126

View Full Product Details