Assay Design
Gene Transcripts
- Gene Symbol
- hrpf-1
- Entrez Gene ID
-
172395
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_W02D3.11
- Chromosome Location
-
Chr.I: 6753219 - 6756354 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_170888.6 |
NP_740877.1 |
5 - 6 |
1305 |
62 |
View Full Product Details