Assay Design



Gene Transcripts

Gene Symbol
hmr-1
Entrez Gene ID
173007
Gene Name
protein-coding
Gene Aliases
CELE_W02B9.1
Chromosome Location
Chr.I: 11075755 - 11134667 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026479.4 NP_001021650.1 19 - 20 4953 72
GenBank mRNA AJ307058.1 - 19 - 20 4953 72

View Full Product Details