Assay Design



Gene Transcripts

Gene Symbol
ced-1
Entrez Gene ID
173064
Gene Name
protein-coding
Gene Aliases
CELE_Y47H9C.4
Chromosome Location
Chr.I: 11851511 - 11871864 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026601.6 NP_001021772.1 9 - 10 3060 126
NM_170927.5 NP_740922.1 9 - 10 3060 126
GenBank mRNA AF332568.1 - 9 - 10 3054 126

View Full Product Details