Assay Design



Gene Transcripts

Gene Symbol
rig-6
Entrez Gene ID
173828
Gene Name
protein-coding
Gene Aliases
CELE_C33F10.5
Chromosome Location
Chr.II: 4764147 - 4812086 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026843.7 NP_001022014.1 8 - 9 2319 73
NM_001377791.2 NP_001364725.1 2 - 3 1053 73
NR_151554.1 5 - 6 1531 73

View Full Product Details