Assay Design



Gene Transcripts

Gene Symbol
dyf-13
Entrez Gene ID
182970
Gene Name
protein-coding
Gene Aliases
CELE_C27H5.7
Chromosome Location
Chr.II: 7168793 - 7171355 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171019.3 NP_741021.1 5 - 6 623 128
NM_171020.5 NP_741022.1 5 - 6 613 128

View Full Product Details