Assay Design
Gene Transcripts
- Gene Symbol
- arx-4
- Entrez Gene ID
-
175247
- Gene Name
- Probable actin-related protein 2/3 complex subunit 2
- Gene Aliases
-
CELE_Y6D11A.2
- Chromosome Location
-
Chr.III: 862306 - 865398 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001384017.1 |
|
3 - 4 |
230 |
64 |
View Full Product Details