Assay Design



Gene Transcripts

Gene Symbol
ptp-1
Entrez Gene ID
175463
Gene Name
protein-coding
Gene Aliases
CELE_C48D5.2
Chromosome Location
Chr.III: 3590722 - 3611971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065331.4 NP_497732.1 11 - 12 1867 92
NM_065332.4 NP_497733.1 4 - 5 678 92
NM_171099.5 NP_741112.1 4 - 5 511 92

View Full Product Details