Assay Design



Gene Transcripts

Gene Symbol
ptp-1
Entrez Gene ID
175463
Gene Name
protein-coding
Gene Aliases
CELE_C48D5.2
Chromosome Location
Chr.III: 3590722 - 3611971 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065331.4 NP_497732.1 16 - 17 3041 74
NM_065332.4 NP_497733.1 9 - 10 1852 74
NM_171099.5 NP_741112.1 9 - 10 1685 74

View Full Product Details