Assay Design
Gene Transcripts
- Gene Symbol
- ubq-1
- Entrez Gene ID
-
175840
- Gene Name
- Ubiquitin-related
- Gene Aliases
-
CELE_F25B5.4
- Chromosome Location
-
Chr.III: 5940241 - 5943841 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171139.7 |
NP_741157.1 |
3 - 4 |
1010 |
95 |
View Full Product Details